Pharmaceutical secondary standard, L-Glutamic acid, CRM, Supelco®
PHR1107-1G
: Merck
:
- Pk:1 g
- Standards:Reference standard
- CAS No.:56-86-0
- Description:L-Glutamic acid
- Melting point:205 °C (dec.) (lit.)
- Formula:HO₂CCH₂CH₂CH(NH₂)CO₂H
- Description:L-Glutamic acid
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
These secondary standards offer multi-traceability to the USP, EP (Ph. Eur.) and BP primary standards, where they are available.
: Accordance with ISO/IEC 17025:2005 (A Class Cert AT-1467), ISO GUIDES 34:2009 (A Class Cert AR-1470).