Pharmaceutical secondary standard, dimethyl sulphoxide (DMSO), CRM, Supelco®
Catalog # PHR1309-3G
Supplier: Merck
CAS Number:
Specifications
- Pk:3 g
- Standards:Pharmaceutical standard
- CAS No.:67-68-5
- Description:Dimethyl sulphoxide
- Formula:C₂H₆OS
- Description:Dimethyl sulphoxide (DMSO)
Specifications
About this item
This certified pharmaceutical secondary standards for application in quality control provides pharma laboratories and manufacturers with a convenient and cost-effective alternative to pharmacopeia primary standards.
This secondary standard offers multi-traceability to the USP and EP (PhEur) primary standards.
Certifications: Accordance with ISO/IEC 17025:2005 (AClass Cert AT-1467), ISO Guides 34:2009 (AClass Cert AR-1470).