Ammonium sulphate ≥99%, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Fluka™
:7783-20-2
:
:(NH₄)₂SO₄
:MFCD00003391
:132.14 g/mol
:13,00559
:350 °C
:1.760 g/cm³ (20 °C)
:Ambient
:231-984-1
:01-2119455044-46
- Assay:Min. 99.0 %
- Water insoluble matter:Max. 0.005 %
- Residue on ignition (SO₄):Max. 0.005 %
- pH (5 %, 25 °C):5.0 - 6.0
- Arsenic (As):Max. 0.2 ppm
- Calcium (Ca):Max. 0.001 %
- Cadmium (Cd):Max. 1 ppm
- Cobalt (Co):Max. 0.0005 %
- Chromium (Cr):Max. 0.0005 %
- Copper (Cu):Max. 2 ppm
- Iron (Fe):Max. 3 ppm
- Potassium (K):Max. 0.005 %
- Magnesium (Mg):Max. 0.0002 %
- Manganese (Mn):Max. 0.0005 %
- Sodium (Na):Max. 0.002 %
- Nickel (Ni):Max. 0.0005 %
- Lead (Pb):Max. 2 ppm
- Zinc (Zn):Max. 0.0001 %
- Heavy metals (ICP) ACS:Max. 2 ppm
- Heavy metals (as Pb):Max. 2 ppm
- Chloride (Cl):Max. 3 ppm
- Nitrate (NO₃):Max. 0.001 %
- Phosphate (PO₄):Max. 5 ppm
Frequently Bought Together