Order Entry
Slovakia
ContactUsLinkComponent
Dimethyl sulphoxide ≥99.5%, Reagent Grade
Dimethyl sulphoxide ≥99.5%, Reagent Grade
Supplier:  HONEYWELL
   Sustainable Options Available
Certificates
undefined
Dimethyl sulphoxide ≥99.5%, Reagent Grade
Supplier:  HONEYWELL
   Sustainable Options Available

Specifications

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default

Specifications

Specification Test Results

  • Appearance:
    complying
  • Identity (NMR):
    complying
  • Assay (GC):
    Min. 99.5 %
  • Water (Karl Fischer):
    Max. 0.1 %
  • Vapor Density:
    2.7 (vs air)
  • Autoignition Temperature:
    573 °F
  • Vapor Pressure:
    0.42 mmHg ( 20 °C)
  • Explosion Limit:
    42 %, 63 °F
  • Refractive Index:
    n20/D 1.479 (lit.)

Specification Test Results

Related Information

Frequently Bought Together