Order Entry
Slovakia
ContactUsLinkComponent
Total organic carbon (TOC) vial, Nalgene®
Total organic carbon (TOC) vial, Nalgene®
Catalog # NALG1000-0000
   Low Manufacturing Impact
Total organic carbon (TOC) vial, Nalgene®
Catalog # NALG1000-0000
   Low Manufacturing Impact
Specifications
  • Cap included:
    Yes
  • Cap Size:
    33-400
  • Ø×H:
    96×217 mm
  • Capacity:
    1000,00 ml
  • Colour:
    Clear
  • Description:
    Boston round bottle with closure
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    NAR Certificate of Retirement
  • Pk:
    1
Specifications
About this item
TOC-certified vials are low-level–certified. Several sizes of containers are available, including the popular autosampler vials cleaned, certified and ready to use. Simplify and reduce the cost of cleaning validations.
  • Low background is perfect for preparation and storage of standards
  • Off-line and grab sampling of high-purity water