Order Entry
Slovakia
ContactUsLinkComponent
Perchloric acid 0.1 mol/l (0.1 N) in anhydrous acetic acid, AVS TITRINORM volumetric solution
Flammable
Corrosive
Perchloric acid 0.1 mol/l (0.1 N) in anhydrous acetic acid, AVS TITRINORM volumetric solution
Catalog # VWRS30110.363
Supplier:  VWR Collection
CAS Number:  
undefined
Flammable
Corrosive
Perchloric acid 0.1 mol/l (0.1 N) in anhydrous acetic acid, AVS TITRINORM volumetric solution
Catalog # VWRS30110.363
Supplier:  VWR Collection
CAS Number:  

Specifications

  • Pack type:
    Plastic container
  • Pk:
    5 L
  • Seq:
    5190

Specifications

Specification Test Results

  • Titer (20°C; real value 0.2 % accuracy):
    0.0998 - 0.1002 mol/l
  • NIST traceable:
    Confirmed

Specification Test Results

About this item

 :
 :100.46 g/mol
Boiling Pt:120 °C (1013 hPa)
Density:1.050 g/cm³ (20 °C)
Flash Pt:40 °C (closed cup)
Storage Temperature:Ambient
EINECS:231-512-4
UN:2920
ADR:8 (3),II
 :GHS02
 :GHS05
CAS Number:7601-90-3
Formula:HClO₄
MDL Number:MFCD00011325