Order Entry
Slovakia
ContactUsLinkComponent
 
 
 
(±)-2-Butanol ≥98%, GPR RECTAPUR®
  VWRS20811.550
 :  
undefined
 
 
(±)-2-Butanol ≥98%, GPR RECTAPUR®
  VWRS20811.550
 :  

 

  • Pk:
    200 L
  • Seq:
    5549

 

 

  • Assay:
    Min. 98 %
  • Boiling point:
    98.5 - 100.5 °C
  • Density (20/4):
    0.805 - 0.809
  • Evaporation residue:
    Max. 100 ppm

 

 

 :
 :74.12 g/mol
 :13,01540
 :94 °C (1013 hPa)
 :–115 °C
 :0.8534 g/cm³ (20 °C)
 :23 °C
 :Ambient
 :201-158-5
 :1120
 :3,III
 :GHS02
 :GHS07
 :78-92-2
 :H₃CCH₂CH(OH)CH₃
 :MFCD00004569