Order Entry
Slovakia
Orders LinkContactUsLinkComponent
Phenanthrene

Phenanthrene

1
Catalog # 48569.
Irritant
Environment
Supplier:   Merck
Phenanthrene
Catalog # 48569.
Supplier:  Merck
CAS Number:  85-01-8
Phenanthrene
Phenanthrene

1

CAS Number:  85-01-8
Price
148,07 €
Log in for availability
1 * 1 items
Product Details & Documents
  • CAS Number:85-01-8
  • Formula:C₁₄H₁₀
  • MDL Number:MFCD00001168
  • Molecular Weight:178.23 g/mol
  • Merck Index:12,07354
  • Boiling Pt:340 °C (1013 hPa)
  • Melting Pt:97…99 °C
  • Density:1.19 g/cm³ (20 °C)
  • Flash Pt:153 °C
  • Storage Temperature:Ambient
  • EINECS:201-581-5
  • UN:3077
  • ADR:9,III
Specifications

Specifications

  • Pk:
    1

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.