Order Entry
Slovakia
Orders LinkContactUsLinkComponent
Phenanthrene tech. 90%, Technical Grade

Phenanthrene tech. 90%, Technical Grade

Irritant
Environment
Phenanthrene tech. 90%, Technical Grade
Certificates
Phenanthrene tech. 90%, Technical Grade
Phenanthrene tech. 90%, Technical Grade

3 Options Available Below

Product Details & Documents

Purity: 90% CAS No.: 85-01-8 Molecular Formula: C14H10 Formula Weight: 178.23 Flash Point: 171°(339°F) Melting Point: 97-101° Boiling Point: 340° Solubility: Soluble in alcohol, benzene, toluene, and glacial acetic acid Beilstein Reference: 1905428 Merck Reference: 14,7212 UN No.: 3077 MDL No.: MFCD00001168

  • CAS Number:85-01-8
  • Formula:C₁₄H₁₀
  • MDL Number:MFCD00001168
  • Molecular Weight:178.23 g/mol
  • Merck Index:14,7212
  • Boiling Pt:340 °C
  • Melting Pt:97…101 °C
  • Density:0.980
  • Flash Pt:171 °C (339 °F)
  • Storage Temperature:Ambient
  • EINECS:201-581-5
  • UN:3077
  • ADR:9,III
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.