


Mercury (II) oxide, yellow ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:21908-53-2
:
Formula:HgO
MDL Number:MFCD00011045
:216.59 g/mol
Merck Index:13,05908
Melting Pt:500 °C
EINECS:244-654-7
UN:1641
ADR:6.1,II
:GHS06
:GHS08
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Appearance:Yellow to light orange Powder
- Titration Complexometric:≥99.0 %
- Chloride (Cl):≤250 ppm
- Sulfate (SO4):≤100 ppm
- Iron (Fe):≤30 ppm
- Insoluble matter:≤0.03 % (in dilute HCl)
- Residue after reduction:≤0.05 %
- Nitrogen compounds (as N):≤0.005 %
Specification Test Results
Frequently Bought Together