

Copper(II) sulphate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:7758-98-7
:
Formula:CuSO₄
MDL Number:MFCD00010981
:159.61 g/mol
Merck Index:12,02722
Melting Pt:200 °C
Density:3.603 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:231-847-6
ADR:6.1,III
:M-Clarity™ quality level = MQ200
UN:3288
:GHS07
:GHS09
Frequently Bought Together