Copper(II) chloride, anhydrous 99%, extra pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Copper dichloride, Cupric chloride
CAS Number:7447-39-4
:
Formula:Cl₂Cu
MDL Number:MFCD00010972
Boiling Pt:993 °C (760 mmHg)
Molecular Weight:134.45 g/mol
Melting Pt:498 °C
ADR:8 III
Merck Index:13,02660
EINECS:231-210-2
UN:2802
:GHS05
:GHS07
:GHS09
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Brown Crystalline powder
- Assay:≥98.5 % (CuCl2)
- Water:≤0.75 %
- Impurity:≤0.75 % (Cupric nitrate)
- Insoluble matter:≤0.25 % (in water)
Specification Test Results
Frequently Bought Together