Dimethyl sulphoxide ≥99.7%, Hybri-Max™, sterile-filtered, Sigma-Aldrich®
SIALD2650-100ML
: Merck
:
- Pack type:Glass bottle
- Pk:100 mL
- Seq:2440
:
:78.14 g/mol
:13,03285
:189 °C (1013 hPa)
:18.5 °C
:1.101 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ300
:87 °C
:Ambient
:200-664-3
:67-68-5
:(CH₃)₂SO
:MFCD00002089