Dimethyl sulphoxide, tested according to Ph. Eur., Riedel-de Haën™
39291-100ML
:
- Pack type:Glass bottle
- Pk:100 mL
- Seq:2440
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default
- Identity (IR):complying
- Density (D 20/20):1.100 - 1.104
- Refractive index (n 20/D):1.478 - 1.480
- Congealing - Freezing Point:Min. 18.3 °C
- Water (Karl Fischer):Max. 0.2 %
- Acidity:complying
- Absorbance at 295 nm:Max. 0.20
- Absorbance at 285 nm:Max. 0.20
- Absorbance at 275 nm:Max. 0.30
- Absorption curve:complying
- Related substances (GC):complying
- Residual solvents:complying
:
:78.14 g/mol
:13,03285
:189 °C
:18.5 °C
:1.104 g/cm³ (20 °C)
:28.5 %(V)
:Ambient
:200-664-3
:67-68-5
:(CH₃)₂SO
:MFCD00002089