Order Entry
Slovakia
ContactUsLinkComponent
Dimethyl sulphoxide, extra dry 99.7+%, AcroSeal®
Dimethyl sulphoxide, extra dry 99.7+%, AcroSeal®
   Sustainable Options Available
Certificates
undefined
Dimethyl sulphoxide, extra dry 99.7+%, AcroSeal®
   Sustainable Options Available
Specifications
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default
Specifications
Specification Test Results
  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Water:
    ≤0.005 %
  • GC:
    ≥99.70 %
  • Residue after evaporation:
    ≤0.001 %