Order Entry
Slovakia
ContactUsLinkComponent
Ammonium dihydrogen phosphate ≥98% ACS, Fluka™
Ammonium dihydrogen phosphate ≥98% ACS, Fluka™
Supplier:  HONEYWELL
Certificates
Ammonium dihydrogen phosphate ≥98% ACS, Fluka™
Supplier:  HONEYWELL
Certificates

Specification Test Results

  • Identity (IR):
    complying
  • Assay:
    Min. 98.0 %
  • Water insoluble matter:
    Max. 0.005 %
  • pH (5 %, 25 °C):
    3.8 - 4.4
  • Calcium (Ca):
    Max. 0.001 %
  • Iron (Fe):
    Max. 0.001 %
  • Potassium (K):
    Max. 0.005 %
  • Magnesium (Mg):
    Max. 5 ppm
  • Sodium (Na):
    Max. 0.005 %
  • Heavy metals (as Pb):
    Max. 5 ppm
  • Chloride (Cl):
    Max. 5 ppm
  • Nitrate (NO₃):
    Max. 0.001 %
  • Sulfate (SO₄):
    Max. 0.01 %

Specification Test Results

Related Information

Frequently Bought Together