Ammonium dihydrogen phosphate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:7722-76-1
:
Formula:NH₄H₂PO₄
MDL Number:MFCD00003396
Merck Index:13,00547
Melting Pt:190 °C
Density:1.8 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
:115.03 g/mol
Storage Temperature:Ambient
EINECS:231-764-5
Frequently Bought Together