Ammonium dihydrogen phosphate ≥99.995% (metals basis), Puratronic®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7722-76-1
:
Formula:NH₄H₂PO₄
MDL Number:MFCD00003396
Merck Index:14,543
Melting Pt:190 °C
Density:1.803
:115.03 g/mol
EINECS:231-764-5
:GHS07
Storage Temperature:Ambient
Frequently Bought Together