Order Entry
Slovakia
ContactUsLinkComponent
Ammonium fluoride ≥98.0%, AnalaR NORMAPUR® analytical reagent
Toxic
Ammonium fluoride ≥98.0%, AnalaR NORMAPUR® analytical reagent
Supplier:  VWR Chemicals
Certificates
undefined
Toxic
Ammonium fluoride ≥98.0%, AnalaR NORMAPUR® analytical reagent
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 98.0 %
  • Ammonium hydrogen difluoride:
    Max. 1 %
  • Heavy metals (as Pb):
    Max. 5 ppm
  • Water:
    Max. 5 %
  • Cl (Chloride):
    Max. 10 ppm
  • SiF6 (Hexafluorosilicate):
    Max. 0.1 %
  • SO4 (Sulphate):
    Max. 50 ppm
  • Fe (Iron):
    Max. 5 ppm
  • K (Potassium):
    Max. 30 ppm
  • Na (Sodium):
    Max. 80 ppm

Specification Test Results

Frequently Bought Together