
Triethyl phosphate ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:78-40-0
:
Formula:C₆H₁₅O₄P
MDL Number:MFCD00009077
:182.16 g/mol
Merck Index:14,9674
Boiling Pt:215 °C
Melting Pt:–57 °C
Density:1.072
Flash Pt:115 °C (240 °F)
EINECS:201-114-5
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together