Order Entry
Slovakia
ContactUsLinkComponent
Barium sulphate, Sigma-Aldrich®
Barium sulphate, Sigma-Aldrich®
Catalog # SIAL202762-20G
Supplier:  Merck
CAS Number:  
Barium sulphate, Sigma-Aldrich®
Catalog # SIAL202762-20G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    20 g
  • Seq:
    3290

Specifications

About this item

 :
 :233.39 g/mol
Merck Index:13,00997
Boiling Pt:1600 °C (1013 hPa)
Melting Pt:1345 °C
Density:4.5 g/cm³ (25 °C)
 :M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:231-784-4
CAS Number:7727-43-7
Formula:BaSO₄
MDL Number:MFCD00003455