Order Entry
Slovakia
ContactUsLinkComponent
Magnesium hydroxide carbonate light, Sigma-Aldrich®
Magnesium hydroxide carbonate light, Sigma-Aldrich®
Catalog # SIAL13118-1KG
Supplier:  Merck
CAS Number:  
Magnesium hydroxide carbonate light, Sigma-Aldrich®
Catalog # SIAL13118-1KG
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
 :84.31 g/mol
Density:2.16 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:235-192-7
CAS Number:12125-28-9
 :MFCD00149783
Formula:MgCO₃ (linear)