
Barium chloride, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:10361-37-2
:
Formula:BaCl₂
MDL Number:MFCD00003445
:208.24 g/mol
Merck Index:12,00998
Boiling Pt:1560 °C (1013 hPa)
Melting Pt:∼963 °C
Density:2.33 g/cm³ (20 °C)
UN:1564
ADR:6.1,III
:M-Clarity™ quality level = MQ100
EINECS:233-788-1
:GHS06
Storage Temperature:Ambient
Frequently Bought Together