Sodium orthovanadate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:13721-39-6
:
Formula:Na₃O₄V
MDL Number:MFCD00003511
:183.91 g/mol
Melting Pt:850…866 °C
:M-Clarity™ quality level = MQ200
EINECS:237-287-9
:GHS07
Storage Temperature:Ambient
Frequently Bought Together