

Potassium thiocyanate, Supelco®
Catalog # 1.05124.1000
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 kg
- Seq:4024
Specifications
About this item
:
Merck Index:12,07866
Boiling Pt:500 °C (1013 hPa)
Melting Pt:175 °C
Density:1.61 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
:97.18 g/mol
EINECS:206-370-1
:GHS05
:GHS07
Storage Temperature:Ambient
CAS Number:333-20-0
Formula:KSCN
MDL Number:MFCD00011413