Potassium hydroxide ≥85%, pellets ACS, Fluka™
Supplier: HONEYWELL
Certificates
About this item
CAS Number:1310-58-3
:
Formula:KOH
MDL Number:MFCD00003553
:56.11 g/mol
Merck Index:12,07806
Boiling Pt:1.327 °C (1.013 hPa)
Melting Pt:360 °C
Density:2.04 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-181-3
UN:1813
ADR:8,II
REACH:01-2119487136-33
:GHS07
:GHS05
Specification Test Results
- Assay:Min. 85.0 %
- Assay of K₂CO₃:Max. 2.0 %
- Calcium (Ca):Max. 0.005 %
- Iron (Fe):Max. 0.001 %
- Magnesium (Mg):Max. 0.002 %
- Sodium (Na):Max. 0.05 %
- Nickel (Ni):Max. 0.001 %
- Form:pellets
- Heavy metals (ICP) ACS:Max. 0.001 %
- Chloride (Cl):Max. 0.01 %
- Phosphate (PO₄):Max. 5 ppm
- Sulfate (SO₄):Max. 0.003 %
- Total N:Max. 0.001 %
- Appearance:complying
Specification Test Results
Frequently Bought Together