Potassium hydroxide ca. 85%, pellets ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1310-58-3
:
Formula:KOH
MDL Number:MFCD00003553
:56.11 g/mol
Merck Index:12,07806
Boiling Pt:1320 °C (760 mmHg)
Melting Pt:360 °C
Density:2,04 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-181-3
UN:1813
ADR:8,II
REACH:01-2119487136-33
:GHS07
:GHS05
Specification Test Results
- Appearance:White Pellets
- Titration with HCl:≥85 % (KOH)
- Chloride (Cl):≤0.01 %
- Phosphate (PO4):≤5 ppm
- Sulfate (SO4):≤0.003 %
- Potassium carbonate:≤2.0 % (K2CO3)
- Calcium (Ca):≤0.005 %
- Iron (Fe):≤0.001 %
- Magnesium (Mg):≤0.002 %
- Nickel (Ni):≤0.001 %
- Sodium (Na):≤0.05 %
- Heavy metals (ICP-OES):≤0.001 %
- Nitrogen compounds (as N):≤0.001 %
Specification Test Results
Frequently Bought Together