Order Entry
Slovakia
ContactUsLinkComponent
Sodium chloride ≥99%, Reagent Grade
Sodium chloride ≥99%, Reagent Grade
Supplier:  HONEYWELL
Certificates
Sodium chloride ≥99%, Reagent Grade
Supplier:  HONEYWELL
Certificates

Specification Test Results

  • Appearance:
    complying
  • Assay (argentometric):
    99.5 - 100.5 %
  • Colour of the solution:
    complying
  • Turbidity of solution:
    complying

Specification Test Results

Related Information

Frequently Bought Together