

Periodic acid, Sigma-Aldrich®
Catalog # SIAL379891-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:227.94 g/mol
Merck Index:13,07248
Melting Pt:122 °C
Density:1.3875 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:233-937-0
UN:3085
ADR:5.1,I
:M-Clarity™ quality level = MQ200
:GHS03
:GHS05
CAS Number:10450-60-9
Formula:H₅IO₆
MDL Number:MFCD00011344