

Iodic acid ≥99.5% ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7782-68-5
:
Formula:HIO₃
MDL Number:MFCD00011348
:175.91 g/mol
Merck Index:14,5012
Melting Pt:110 °C (dec.)
Density:4.629
Storage Temperature:Ambient
EINECS:231-962-1
UN:3085
ADR:5.1,II
:GHS03
:GHS05
Related Information
Frequently Bought Together