Fluorescein disodium salt, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:518-47-8
:
Formula:C₂₀H₁₀Na₂O₅
MDL Number:MFCD00167039
:376.28 g/mol
Merck Index:13,04185
Melting Pt:> 300 °C
Density:1.601 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:208-253-0
Frequently Bought Together