Order Entry
Slovakia
ContactUsLinkComponent
Fluorescein disodium salt Reag. Ph. Eur., Supelco®
Fluorescein disodium salt Reag. Ph. Eur., Supelco®
Supplier:  Merck
Certificates
undefined
Fluorescein disodium salt Reag. Ph. Eur., Supelco®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    500 g/l (20 °C)
  • Melting Point:
    300 °C
  • Molar Mass:
    376.27 g/mol
  • Bulk Density:
    600 kg/m3
  • pH:
    8.3 (10 g/l, H2O, 20 °C)

Specification Test Results

Related Information

Frequently Bought Together