Piperazine 99%, extra pure
:110-85-0
:
:C₄H₁₀N₂
:MFCD00005953
:86.14 g/mol
:13,07545
:146…148 °C (760 mmHg)
:107…111 °C
:1,1 g/cm³ (20 °C)
:65 °C
:Ambient
:203-808-3
:2579
:8,III
:GHS08
:GHS05
:GHS02
- Appearance:White to light yellow Crystalline flakes
- Infrared spectrum:Conforms
- Water:≤1 %
- GC:≥98.5 %
- Color of solution:≤30 APHA (50% in methanol)
Frequently Bought Together