α-Terpinene 90% tech.
: p-Mentha-1,3-diene, 1-Isopropyl-4-methylcyclohexa-1,3-diene
- :99-86-5
- :C₁₀H₁₆
- :MFCD00001534
- :136.24 g/mol
- :173 °C (1013 hPa)
- :–48 °C
- :0.837 g/cm³
- :46 °C
- :Freezer
- :202-795-1
- :2319
- :3,III
: p-Mentha-1,3-diene, 1-Isopropyl-4-methylcyclohexa-1,3-diene
- :99-86-5
- :C₁₀H₁₆
- :MFCD00001534
- :136.24 g/mol
- :173 °C (1013 hPa)
- :–48 °C
- :0.837 g/cm³
- :46 °C
- :Freezer
- :202-795-1
- :2319
- :3,III
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4770 to 1.4790 (20°C, 589 nm)
- Density:(20°C) 0.832 to 0.842 g/ml
- GC:≥88 %
Product Family Options
- Pack typePk
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



