
Benzotriazole ≥99.0% for amperometric det. of Cu, Fluka™
Supplier: HONEYWELL
Certificates
About this item
CAS Number:95-14-7
:
Formula:C₆H₅N₃
MDL Number:MFCD00005699
:119.13 g/mol
Merck Index:13,01109
Boiling Pt:350 °C (1013 hPa)
Melting Pt:95…99 °C
Density:1,34 g/cm³ (20 °C)
Flash Pt:170 °C (closed cup)
Storage Temperature:Ambient
EINECS:202-394-1
UN:3077
ADR:9,III
:GHS07
Specification Test Results
- Identity (IR):complying
- Assay (HPLC):Min. 99.0 %
- Melting point:95 - 99 °C
- Residue on ignition:Max. 0.1 %
Specification Test Results
Frequently Bought Together