Supelco® Organic Reference Standard, p-Terphenyl
Catalog # 45802-250MG
Supplier: Merck
CAS Number:
Specifications
- Pk:250 mg
- Standards:Analytical standard
- CAS No.:92-94-4
- Description:p-Terphenyl
- Storage conditions:2...30 °C
- Hazard class description:H319 - H413
- Shipping conditions:STD
- MDL No.:MFCD00003061
- M-Clarity quality level:MQ100
- Synonym(s):1,4-Diphenylbenzene; PT; PTP
- Melting point:212 - 213 °C (lit.)
- Formula:C₆H₅C₆H₄C₆H₅
- Molecular weight:230,30
- Flash point:207 °C - Closed cup
- Boiling point:389 °C (lit.)
Specifications
About this item
- GHS signal: GHS07, GHS09
Caution: P264 - P273 - P280 - P305 + P351 + P338 - P337 + P313 - P501