Supelco® Organic Reference Standard, 1,2,3-Trimethylbenzene
Catalog # 45935-250MG
Supplier: Merck
CAS Number:
Specifications
- Pk:250 mg
- Standards:Analytical standard
- CAS No.:526-73-8
- Description:1,2,3-Trimethylbenzene
- Storage conditions:2...30 °C
- Hazard class description:H226 - H315 - H319 - H335
- Shipping conditions:STD
- MDL No.:MFCD00008520
- M-Clarity quality level:MQ100
- Synonym(s):Hemellitol
- Melting point:–25 °C (lit.)
- Formula:C₆H₃(CH₃)₃
- Molecular weight:120,19
- Density:0,894 g/ml at 25 °C (lit.)
- Flash point:53 °C - Closed cup
- Boiling point:175 - 176 °C (lit.)
Specifications
About this item
- GHS signal: GHS02, GHS07
Caution: P210 - P302 + P352 - P305 + P351 + P338