Order Entry
Slovakia
ContactUsLinkComponent
Supelco® Organic Reference Standard, m-Cymene
Supelco® Organic Reference Standard, m-Cymene
  30037-250MG
 :  Merck
 :  
undefined
Supelco® Organic Reference Standard, m-Cymene
  30037-250MG
 :  Merck
 :  

 

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 mg
  • Standards:
    Analytical standard
  • CAS No.:
    535-77-3
  • Description:
    m-Cymene
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H226
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00008891
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    1-Isopropyl-3-methylbenzene; 3-Isopropyltoluene
  • Melting point:
    –64 °C (lit.)
  • Formula:
    CH₃C₆H₄CH(CH₃)₂
  • Molecular weight:
    134,22
  • Density:
    0,861 g/ml at 25 °C (lit.)
  • Flash point:
    47 °C - closed cup
  • Boiling point:
    175 °C (lit.)

 

 

  • GHS signal: GHS02
 : P210 - P233 - P240 - P241 - P242 - P243