
p-Tolyl isothiocyanate, Sigma-Aldrich®
Catalog # SIAL253715-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:149.22 g/mol
Boiling Pt:237 °C (1013 hPa)
Melting Pt:27…30 °C
Storage Temperature:Ambient
ADR:8,III
:M-Clarity™ quality level = MQ200
EINECS:210-745-5
UN:2923
:GHS07
CAS Number:622-59-3
Formula:C₈H₇NS
MDL Number:MFCD00004813