
3,4-Difluorobenzylamine, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:72235-53-1
:
Formula:C₇H₇F₂N
MDL Number:MFCD00010145
:143.14 g/mol
Boiling Pt:174 °C (1013 hPa)
Melting Pt:31 °C
Density:1.21 g/cm³ (20 °C)
EINECS:276-503-6
ADR:8,III
:M-Clarity™ quality level = MQ200
UN:2735
:GHS05
Frequently Bought Together