Order Entry
Slovakia
ContactUsLinkComponent
 
Supelco® PESTANAL® Organic Reference Standard, Eugenol
  35995-250MG
 :  Merck
 :  
undefined
Supelco® PESTANAL® Organic Reference Standard, Eugenol
  35995-250MG
 :  Merck
 :  

 

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 mg
  • Standards:
    PESTANAL®, Analytical standard
  • CAS No.:
    97-53-0
  • Description:
    Eugenol
  • Storage conditions:
    RT
  • Hazard class description:
    H317 - H319
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00008654
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    2-Methoxy-4-(2-propenyl)phenol; 4-Allyl-2-methoxyphenol; 4-Allylguaiacol
  • Melting point:
    –12…+10 °C (lit.)
  • Formula:
    4-(H₂C=CHCH₂)C₆H₃-2-(OCH₃)OH
  • Molecular weight:
    164,20
  • Density:
    1,067 g/ml at 25 °C (lit.)
  • Flash point:
    112 °C - closed cup
  • Boiling point:
    254 °C (lit.)

 

 

  • GHS signal: GHS07
 : P261 - P264 - P272 - P280 - P302 + P352 - P305 + P351 + P338