3,5-Dinitrosalicylic acid, Sigma-Aldrich®
Catalog # SIAL128848-500G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
- Seq:3645
Specifications
About this item
:
:228.12 g/mol
Melting Pt:170…174 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:M-Clarity™ quality level = MQ200
EINECS:210-204-3
:GHS07
CAS Number:609-99-4
Formula:C₇H₄N₂O₇
MDL Number:MFCD00007104