Dimethyl phthalate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:131-11-3
:
Formula:C₁₀H₁₀O₄
MDL Number:MFCD00008425
:194.19 g/mol
Merck Index:13,03281
Boiling Pt:283.7 °C (1013 hPa)
Melting Pt:6 °C
Density:1.192 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Flash Pt:146 °C
Storage Temperature:Ambient
EINECS:205-011-6
Frequently Bought Together