Dimethyl phthalate for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:131-11-3
:
Formula:C₁₀H₁₀O₄
MDL Number:MFCD00008425
:194.19 g/mol
Merck Index:13,03281
Boiling Pt:283.7 °C (1013 hPa)
Melting Pt:6 °C
Density:1.192 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Flash Pt:146 °C
Storage Temperature:Ambient
EINECS:205-011-6
Specification Test Results
- Solubility:- 0.1 g/l (20 °C)
- Melting Point:6 °C
- Molar Mass:194.18 g/mol
- Boiling Point:283 °C (1013 hPa)
- Vapor Pressure:0.008 hPa (20 °C)
- Flash Point:150 °C
- Refractive Index:1.5137 (25 °C, 589 nm)
- Density:1.19 g/cm3 (20 °C)
- pH:7 (H2O, 20 °C)
- Ignition Point:460 °C DIN 51794
Specification Test Results
Related Information
Frequently Bought Together