Order Entry
Slovakia
ContactUsLinkComponent
 
Health
Environment
Dibutyl phthalate, Supelco®
Catalog # 36736-1G
Supplier:  Merck
CAS Number:  
undefined
Health
Environment
Dibutyl phthalate, Supelco®
Catalog # 36736-1G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :278.35 g/mol
Merck Index:13,01586
Boiling Pt:340 °C (1013 hPa)
Melting Pt:–35 °C
Density:1.04 g/cm³ (20 °C)
Flash Pt:171 °C (closed cup)
Storage Temperature:Ambient
EINECS:201-557-4
UN:3082
ADR:9,III
 :M-Clarity™ quality level = MQ100
REACH:01-2119493042-44
 :GHS08
 :GHS09
CAS Number:84-74-2
Formula:C₁₆H₂₂O₄
MDL Number:MFCD00009441