Ethyl benzoate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:93-89-0
:
Formula:C₉H₁₀O₂
MDL Number:MFCD00009109
:150.18 g/mol
Merck Index:13,03802
Boiling Pt:415 °C (1013 hPa)
Melting Pt:–35 °C
Density:1.05 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ300
Flash Pt:91 °C
EINECS:202-284-3
:GHS07
Storage Temperature:Ambient
Frequently Bought Together