Order Entry
Slovakia
ContactUsLinkComponent
2-Chloro-4-nitrobenzamide
2-Chloro-4-nitrobenzamide
Catalog # EHERC10049000
Supplier:  EHRENSTORFER
CAS Number:  
2-Chloro-4-nitrobenzamide
Catalog # EHERC10049000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,1 g
  • Seq:
    3020

Specifications

About this item

 :
 :200.58 g/mol
EINECS:221-143-7
CAS Number:3011-89-0
Formula:C₇H₅ClN₂O₃
MDL Number:MFCD00017119