Pyrrolidine, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:123-75-1
:
Formula:C₄H₉N
MDL Number:MFCD00005249
:71.12 g/mol
Merck Index:12,08199
Boiling Pt:87 °C (1013 hPa)
Melting Pt:–63 °C
Density:0.86 g/cm³ (20 °C)
EINECS:204-648-7
UN:1922
ADR:3,II
:M-Clarity™ quality level = MQ200
Flash Pt:3 °C
:GHS02
:GHS05
:GHS07
Storage Temperature:Ambient
Frequently Bought Together