Brefeldin A 10 mg/ml in DMSO (from Penicillium brefeldianum) ≥98.00% (by HPLC) ready-to-use, Sigma-Aldrich®
SIALB5936-200UL
: Merck
:
- Pk:200 µl
- Seq:1358
- Concentration:10.000 - 10.500 mg/ml
- Purity (HPLC):≥98.00%
- Vapor Pressure:0.56 hPa ( 20 °C)
- Autoignition Temperature:301 °C
- Antibiotic Activity Spectrum :Neoplastics
:
:280.4 g/mol
:M-Clarity™ quality level = MQ200
:Refrigerator
:20350-15-6
:C₁₆H₂₄O₄
:MFCD00083258