1-Hexene, Sigma-Aldrich®
SIAL320323-1L
: Merck
:
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
:
:84.16 g/mol
:63 °C (1013 hPa)
:–140 °C
:0.675 g/cm³ (20 °C)
:–26 °C
:Ambient
:209-753-1
:2370
:3,II
:M-Clarity™ quality level = MQ100
:GHS02
:GHS08
:592-41-6
:C₆H₁₂
:MFCD00009505